Phenoxyacetyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Phenoxyacetyl chloride
M-Clarity™ quality level = MQ100
158623-10G
158623-250G
158623-50G
SIAL158623-10GEA
797
CZK
SIAL158623-10G
SIAL158623-250G
SIAL158623-50G
Phenoxyacetyl chloride, Sigma-Aldrich®
Phenoxyacetyl chloride
Vzorec:
C₈H₇ClO₂ Bod varu: 225 °C (1013 hPa) Bod tání: 28…30 °C Hustota: 1,23 g/cm³ (20 °C) |
MDL Number:
MFCD00000726 CAS číslo: 701-99-5 EINECS: 211-862-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...