Tisk…

Pelargonaldehyde stabilised, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Nonanal, Nonylaldehyde, Nonanal, Nonaldehyde

M-Clarity™ quality level = MQ300

W278203-1KG-K W278203-20KG-K W278203-4KG-K
SIALW278203-1KG-KEA 3520 CZK
SIALW278203-1KG-K SIALW278203-20KG-K SIALW278203-4KG-K
Pelargonaldehyde stabilised, Sigma-Aldrich®
Pelargonaldehyde
Vzorec: C₉H₁₈O
Bod varu: 190 °C (1013 hPa)
Bod tání: –40 °C
Hustota: 0,83 g/cm³ (20 °C)
Bod vzplanutí: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007030
CAS číslo: 124-19-6
EINECS: 204-688-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR