Heptafluorobutyric anhydride (HFAA), Supelco®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
HFBA, 2,2,3,3,4,4,4-Heptafluorobutanoic anhydride, Perfluorobutyric anhydride, HFAA
M-Clarity™ quality level = MQ100
77253-10ML
77253-50ML
77253-10X1ML
77253-10MLEA
5930
CZK
77253-10ML
77253-50ML
77253-10X1ML
Heptafluorobutyric anhydride (HFAA), Supelco®
Heptafluorobutyric anhydride
Vzorec:
C₈F₁₄O₃ MW: 410,06 g/mol Bod varu: 108 °C (1013 hPa) Bod tání: 14 °C Hustota: 1,653 g/cm³ (25 °C) |
MDL Number:
MFCD00000432 CAS číslo: 336-59-4 EINECS: 206-410-8 UN: 3625 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...