Borane dimethyl sulphide complex 2 mol/l in THF, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Borane-methyl sulphide complex
M-Clarity™ quality level = MQ200
192120-100ML
192120-800ML
SIAL192120-100MLEA
3070
CZK
SIAL192120-100ML
SIAL192120-800ML
Borane dimethyl sulphide complex 2 mol/l in THF, Sigma-Aldrich®
Borane dimethyl sulphide complex
Vzorec:
C₂H₉BS MW: 75,97 g/mol |
MDL Number:
MFCD00013189 CAS číslo: 13292-87-0 EINECS: 236-313-6 UN: 3399 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...