Tisk…

Borane dimethyl sulphide complex, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Borane-methyl sulphide complex

M-Clarity™ quality level = MQ200

179825-100ML 179825-25ML 179825-800ML
SIAL179825-100MLEA 7400 CZK
SIAL179825-100ML SIAL179825-25ML SIAL179825-800ML
Borane dimethyl sulphide complex, Sigma-Aldrich®
Borane dimethyl sulphide complex
Vzorec: C₂H₉BS
Bod tání: –27 °C
Hustota: 1,28 g/cm³ (20 °C)
MDL Number: MFCD00013189
CAS číslo: 13292-87-0
EINECS: 236-313-6
UN: 2924
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR