Barium hydroxide hydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
450170-25G
450170-5G
SIAL450170-25GEA
5860
CZK
SIAL450170-25G
SIAL450170-5G
Barium hydroxide hydrate, Sigma-Aldrich®
Barium hydroxide hydrate
Vzorec:
H₂BaO₂·H₂O MW: 189,36 g/mol Storage Temperature: Ambient |
CAS číslo:
40226-30-0 EINECS: 241-234-5 UN: 3262 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...