2-Chloro-5-pyridineboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Chloropyridine-5-boronic acid, 2-Chloro-5-pyridinylboronic acid, 6-Chloro-3-pyridinylboronic acid, (6-Chloropyridin-3-yl)boronic acid, 6-Chloropyridine-3-boronic acid
M-Clarity™ quality level = MQ100
637386-1G
637386-5G
SIAL637386-1GEA
2880
CZK
SIAL637386-1G
SIAL637386-5G
2-Chloro-5-pyridineboronic acid, Sigma-Aldrich®
2-Chloro-5-pyridineboronic acid
Vzorec:
C₅H₅BClNO₂ |
MDL Number:
MFCD03094998 CAS číslo: 444120-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...