Tisk…

4-Cholesten-3-one, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one, (+)-4-Cholesten-3-one

M-Clarity™ quality level = MQ100

188174-10G 188174-1G
SIAL188174-10GEA 4150 CZK
SIAL188174-10G SIAL188174-1G
4-Cholesten-3-one, Sigma-Aldrich®
4-Cholesten-3-one
Vzorec: C₂₇H₄₄O
MW: 384,65 g/mol
Bod tání: 81…82 °C
MDL Number: MFCD00003663
CAS číslo: 601-57-0
EINECS: 210-005-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR