Tisk…

β-Alanin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid

M-Clarity™ quality level = MQ300

146064-25G 146064-2KG 146064-500G
SIAL146064-25GEA 1020 CZK
SIAL146064-25G SIAL146064-2KG SIAL146064-500G
β-Alanin, Sigma-Aldrich®
β-Alanin
Vzorec: C₃H₇NO₂
Bod varu: 201 °C (1013 hPa)
Bod tání: 94 °C
Hustota: 1,44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS číslo: 107-95-9
EINECS: 203-536-5
Merck Index: 13,00204

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR