Tisk…

2-Hydroxyisobutyric acid, (max. 2% H₂O) 99% (dry weight)

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Hydroxy-2-methylpropionic acid, α-Hydroxyisobutyric acid, 2-Hydroxy-2-methylpropanoic acid

A13146.14 A13146.22 A13146.36
A13146.14EA 1270 CZK
A13146.14 A13146.22 A13146.36
2-Hydroxyisobutyric acid, (max. 2% H₂O) 99% (dry weight)
2-Hydroxyisobutyric acid

Purity: 99% CAS No.: 594-61-6 Molecular Formula: C4H8O3 Formula Weight: 104.11 Melting Point: 77-80° Boiling Point: 114°/12mm MDL No.: MFCD00004459

Vzorec: C₄H₈O₃
MW: 104,11 g/mol
Bod varu: 114 °C (12 mmHg)
Bod tání: 77…80 °C
Hustota: 1,24
Storage Temperature: Ambient
MDL Number: MFCD00004459
CAS číslo: 594-61-6
EINECS: 209-848-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR