2-Chloro-4-pyridineboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2-Chloropyridin-4-yl)boronic acid, 2-Chloro-4-pyridinylboronic acid, 2-Chloropyridine-4-boronic acid
M-Clarity™ quality level = MQ100
666513-1G
666513-5G
SIAL666513-1GEA
2870
CZK
SIAL666513-1G
SIAL666513-5G
2-Chloro-4-pyridineboronic acid, Sigma-Aldrich®
2-Chloro-4-pyridineboronic acid
Vzorec:
C₅H₅BClNO₂ |
MDL Number:
MFCD03788416 CAS číslo: 458532-96-2 |