Tisk…

2-Bromo-4-pyridineboronic acid 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: (2-Bromopyridin-4-yl)boronic acid, 2-Bromo-4-pyridinylboronic acid, 2-Bromopyridine-4-boronic acid

H27599.MD H27599.03
H27599.MDEA 3270 CZK
H27599.MD H27599.03
2-Bromo-4-pyridineboronic acid 95%
2-Bromo-4-pyridineboronic acid

Purity: 95% CAS No.: 458532-94-0 Molecular Formula: C5H5BBrNO2 Formula Weight: 201.82 MDL No.: MFCD06798244

Vzorec: C₅H₅BBrNO₂
MW: 201,81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798244
CAS číslo: 458532-94-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR