2-Bromoisobutyric acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A15854.14
A15854.22
A15854.36
A15854.14EA
858
CZK
A15854.14
A15854.22
A15854.36
2-Bromoisobutyric acid ≥98%
2-Bromoisobutyric acid
Purity: 98%
CAS No.: 2052-01-9
Molecular Formula: C4H7BrO2
Formula Weight: 167.00
Flash Point:
Vzorec:
C₄H₇BrO₂ Bod varu: 198…200 °C Bod tání: 42…46 °C Hustota: 1,43 g/cm³ (20 °C) Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004168 CAS číslo: 2052-01-9 EINECS: 218-139-2 UN: 3261 ADR: 8,III Merck Index: 14,1421 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...