Tisk…

DL-α-Fenylalanin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

147966-25G 147966-100G
SIAL147966-25GEA 1130 CZK
SIAL147966-25G SIAL147966-100G
DL-α-Fenylalanin, Sigma-Aldrich®
DL-α-Fenylalanin
Vzorec: C₉H₁₁NO₂
Bod varu: 313 °C (1013 hPa)
Bod tání: 173 °C
MDL Number: MFCD00064225
CAS číslo: 150-30-1
EINECS: 205-756-7
Merck Index: 13,07355

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR