Tisk…

2-Aminoisobutyric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid

M-Clarity™ quality level = MQ200

850993-100G 850993-25G
SIAL850993-100GEA 4810 CZK
SIAL850993-100G SIAL850993-25G
2-Aminoisobutyric acid, Sigma-Aldrich®
2-Aminoisobutyric acid
Vzorec: C₄H₉NO₂
MW: 103,12 g/mol
Bod varu: 204 °C (1013 hPa)
Bod tání: 335 °C
Hustota: 1,278 g/cm³ (20 °C)
MDL Number: MFCD00008049
CAS číslo: 62-57-7
EINECS: 200-544-0
Merck Index: 13,00444

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR