Tisk…

4,4'-Dimethoxybenzil ≥98%

Dodavatel: Thermo Fisher Scientific

A10594.06 A10594.14 A10594.22
A10594.06EA 1170 CZK
A10594.06 A10594.14 A10594.22
4,4'-Dimethoxybenzil ≥98%
4,4'-Dimethoxybenzil

Purity: 98+% CAS No.: 1226-42-2 Molecular Formula: C16H14O4 Formula Weight: 270.29 Melting Point: 132-134° Beilstein Reference: 531345

Vzorec: C₁₆H₁₄O₄
Bod varu: 215…219 °C (3 mmHg)
Bod tání: 132…134 °C
Hustota: 1,36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008405
CAS číslo: 1226-42-2
EINECS: 214-960-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR