4-Nitro-m-toluonitrile ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A11301.06
A11301.09
A11301.14
A11301.06EA
2500
CZK
A11301.06
A11301.09
A11301.14
4-Nitro-m-toluonitrile ≥97%
4-Nitro-m-toluonitrile
Purity: 97%
CAS No.: 96784-54-2
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Melting Point: 78-84°
Beilstein Reference: 2443778
UN No.: 3439
MDL No.: MFCD00017615
Vzorec:
C₈H₆N₂O₂ Bod tání: 78…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017615 CAS číslo: 96784-54-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...