2,4,6-Trimethylbenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A11882.06
A11882.14
A11882.06EA
1970
CZK
A11882.06
A11882.14
2,4,6-Trimethylbenzonitrile ≥98%
2,4,6-Trimethylbenzonitrile
Purity: 98%
CAS No.: 2571-52-0
Molecular Formula: C10H11N
Formula Weight: 145.21
Melting Point: 50-55°
Boiling Point: 79-80°/2mm
UN No.: 3439
MDL No.: MFCD00016378
Vzorec:
C₁₀H₁₁N MW: 145,2 g/mol Bod varu: 79…80 °C (2 mmHg) Bod tání: 50…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016378 CAS číslo: 2571-52-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...