1-Butylpyridinium bromide ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
N-butylpyridinium bromide, 1-n-Butylpyridinium bromide
H26995.18
H26995.06
H26995.18EA
6040
CZK
H26995.18
H26995.06
1-Butylpyridinium bromide ≥99%
1-Butylpyridinium bromide
Purity: 99%
CAS No.: 874-80-6
Molecular Formula: C9H14BrN
Formula Weight: 216.12
Melting Point: 105-108°
MDL No.: MFCD00671496
Vzorec:
C₉H₁₄BrN Bod tání: 105…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671496 CAS číslo: 874-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...