Tisk…

4-Amino-3-nitrobenzonitrile 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B23463.03 B23463.06 B23463.14
B23463.03EA 965 CZK
B23463.03 B23463.06 B23463.14
4-Amino-3-nitrobenzonitrile 98%
4-Amino-3-nitrobenzonitrile

Purity: 98% CAS No.: 6393-40-4 Molecular Formula: C7H5N3O2 Formula Weight: 163.14 Melting Point: 159-161° UN No.: 3439 MDL No.: MFCD00013373

Vzorec: C₇H₅N₃O₂
Bod tání: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00013373
CAS číslo: 6393-40-4
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR