Tisk…

4-Amino-3-iodobenzonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Cyano-2-iodoaniline

657123-1G 657123-5G
SIAL657123-1GEA 1400 CZK
SIAL657123-1G SIAL657123-5G
4-Amino-3-iodobenzonitrile, Sigma-Aldrich®
4-Amino-3-iodobenzonitrile
Vzorec: C₇H₅IN₂
MW: 244,03 g/mol
MDL Number: MFCD04039964
CAS číslo: 33348-34-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR