Tisk…

2-Amino-3,5-dichlorobenzonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

642819-1G 642819-5G
SIAL642819-1GEA 1960 CZK
SIAL642819-1G SIAL642819-5G
2-Amino-3,5-dichlorobenzonitrile, Sigma-Aldrich®
2-Amino-3,5-dichlorobenzonitrile
Vzorec: C₇H₄Cl₂N₂
MW: 187,03 g/mol
MDL Number: MFCD00158947
CAS číslo: 36764-94-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR