Tisk…

p-Tolylacetonitrile ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: P-Toluylacetonitrile, 4-Methylbenzyl cyanide, (4-Methylphenyl)acetonitrile

A12169.06 A12169.14
A12169.06EA 680 CZK
A12169.06 A12169.14
p-Tolylacetonitrile ≥98%
p-Tolylacetonitrile

Purity: 98+% CAS No.: 2947-61-7 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 106°(222°F) Refractive Index: 1.5190 Melting Point: 18° Boiling Point: 242-243° Beilstein Reference: 1099645 UN No.: 3276 MDL No.: MFCD00001922

Vzorec: C₉H₉N
MW: 131,18 g/mol
Bod varu: 242…243 °C
Bod tání: 18 °C
Hustota: 0,989
Bod vzplanutí: 106 °C (222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001922
CAS číslo: 2947-61-7
EINECS: 220-963-2
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR