(2-Nitrophenyl)acetonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Nitrobenzyl cyanide
B20190.06
B20190.14
B20190.06EA
1920
CZK
B20190.06
B20190.14
(2-Nitrophenyl)acetonitrile ≥98%
(2-Nitrophenyl)acetonitrile
Purity: 98%
CAS No.: 610-66-2
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Melting Point: 81-85°
Boiling Point: 178°/12mm
UN No.: 3439
MDL No.: MFCD00007183
Vzorec:
C₈H₆N₂O₂ MW: 162,15 g/mol Bod varu: 178 °C (12 mmHg) Bod tání: 81…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007183 CAS číslo: 610-66-2 EINECS: 210-231-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...