o-Tolylacetonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(2-Methylphenyl)acetonitrile, 2-(o-Tolyl)acetonitrile, 2-Methylbenzyl cyanide
A14787.06
A14787.14
A14787.22
A14787.06EA
841
CZK
A14787.06
A14787.14
A14787.22
o-Tolylacetonitrile ≥98%
o-Tolylacetonitrile
Purity: 98+%
CAS No.: 22364-68-7
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point:
Vzorec:
C₉H₉N MW: 131,18 g/mol Bod varu: 211…213 °C Bod tání: 58…59 °C Hustota: 1,010 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001904 CAS číslo: 22364-68-7 EINECS: 244-937-5 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...