D-α-Phenylalanine ≥99%
Dodavatel: Thermo Scientific
Synonyms:
D-Phenylalanine, H-D-Phe-OH, 3-Phenyl-D-alanine, 3-Phenyl-D(+)-alanine
A10572.06
A10572.14
A10572.22
A10572.06EA
1560
CZK
A10572.06
A10572.14
A10572.22
D-α-Phenylalanine ≥99%
D-α-Phenylalanine
Purity: 99%
CAS No.: 673-06-3
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Melting Point: ca 275° dec.
Beilstein Reference: 2804068
Merck Reference: 14,7271
Optical Rotation: +34° (c=2 in water)
MDL No.: MFCD00004270
Vzorec:
C₉H₁₁NO₂ MW: 165,19 g/mol Bod varu: 295 °C (1013 hPa) Bod tání: ∼ 273 °C…276 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00004270 CAS číslo: 673-06-3 EINECS: 211-603-5 Merck Index: 14,7271 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...