(R)-(+)-1,2-Epoxypropane ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B21884.06
B21884.14
B21884.22
B21884.06EA
2610
CZK
B21884.06
B21884.14
B21884.22
(R)-(+)-1,2-Epoxypropane ≥99%
(R)-(+)-1,2-Epoxypropane
Purity: 99%
CAS No.: 15448-47-2
Molecular Formula: C3H6O
Formula Weight: 58.08
Flash Point: -37°(-35°F)
Odour: Ether-like
Boiling Point: 34-35°
Beilstein Reference: 79764
Merck Reference: 14,7856
UN No.: 1280
MDL No.: MFCD00066211
Vzorec:
C₃H₆O Bod varu: 34…35 °C Hustota: 0,829 Bod vzplanutí: –37 °C (–35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00066211 CAS číslo: 15448-47-2 UN: 1280 ADR: 3,I Merck Index: 14,7856 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...