D-Alanine ≥99%
Dodavatel: Thermo Scientific
Synonyms:
(R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
A10231.06
A10231.14
A10231.22
A10231.06EA
1460
CZK
A10231.06
A10231.14
A10231.22
D-Alanine ≥99%
D-Alanine
Purity: 99%
CAS No.: 338-69-2
Molecular Formula: C3H7NO2
Formula Weight: 89.09
Melting Point: ca 295° dec.
Beilstein Reference: 1720249
Merck Reference: 14,204
Optical Rotation: -14° (c=5 in 5N HCl)
MDL No.: MFCD00008077
Vzorec:
C₃H₇NO₂ MW: 89,09 g/mol Bod varu: 189 °C (1013 hPa) Bod tání: ∼ 295 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00008077 CAS číslo: 338-69-2 EINECS: 206-418-1 Merck Index: 14,204 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...