Fumaronitrile, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
131016-25G
131016-5G
SIAL131016-25GEA
10880
CZK
SIAL131016-25G
SIAL131016-5G
Fumaronitrile, Sigma-Aldrich®
Fumaronitrile
Vzorec:
C₄H₂N₂ Bod varu: 186 °C (1013 hPa) Bod tání: 96 °C Hustota: 1,21 g/cm³ (20 °C) |
MDL Number:
MFCD00001928 CAS číslo: 764-42-1 EINECS: 212-122-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...