Phenylacetyl chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
P16753-100G
P16753-500G
P16753-5G
SIALP16753-100GEA
2150
CZK
SIALP16753-100G
SIALP16753-500G
SIALP16753-5G
Phenylacetyl chloride, Sigma-Aldrich®
Phenylacetyl chloride
Vzorec:
C₈H₇ClO MW: 154,6 g/mol Bod varu: 225 °C (1013 hPa) Bod tání: 91 °C Hustota: 1,1682 g/cm³ (20 °C) Bod vzplanutí: 102 °C |
MDL Number:
MFCD00000729 CAS číslo: 103-80-0 EINECS: 203-146-5 UN: 2577 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...