L(-)-Sorbóza ≥98%, white crystalline powder
Dodavatel: MP Biomedicals
Sorbose is a ketose belonging to the group of sugars known as monosaccharides. It has a sweetness that is equivalent to sucrose.The commercial production of vitamin C (ascorbic acid) often begins with sorbose
L-Sorbose is used as a starting reagent for the commercial biosynthesis of ascorbic acid (vitamin C).
Freely soluble in water (50 mg/ml-clear, colorless solution); almost insoluble in alcohol.
Vzorec:
C₆H₁₂O₆ Bod varu: 380 °C (1013 hPa) Bod tání: 158 - 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151097 CAS číslo: 87-79-6 EINECS: 201-771-8 Merck Index: 12,08874 |
Výsledky specifikačního testu
Identity Test | Passes |
Purity (HPLC) | ≥98% |
FTIR | Conforms to Standard |
Melting Point | 159 - 175 °C |
Optical Rotation:(c=2,water) | −38° to −48° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...