Isopentyl hexanoate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W207500-1KG-K
W207500-9KG-K
SIALW207500-1KG-KEA
2980
CZK
SIALW207500-1KG-K
SIALW207500-9KG-K
Isopentyl hexanoate, Sigma-Aldrich®
Isopentyl hexanoate
Vzorec:
C₁₁H₂₂O₂ Bod varu: 226 °C (1013 hPa) Hustota: 0,861 g/cm³ (20 °C) |
MDL Number:
MFCD00027280 CAS číslo: 2198-61-0 EINECS: 218-600-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...