Tisk…

Xylitol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

M-Clarity™ quality level = MQ300

W507930-1KG W507930-5KG W507930-SAMPLE
SIALW507930-1KGEA 3870 CZK
SIALW507930-1KG SIALW507930-5KG SIALW507930-SAMPLE
Xylitol, Sigma-Aldrich®
Xylitol
Vzorec: C₅H₁₂O₅
MW: 152.15
Bod varu: 495
Bod tání: 94
Hustota: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS číslo: 87-99-0
EINECS: 201-788-0
Merck Index: 13,10140

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR