Xylitol 99, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
MQ200
X3375-25G
X3375-100G
X3375-1KG
X3375-500G
X3375-5G
SIALX3375-25GEA
1440
CZK
SIALX3375-25G
SIALX3375-100G
SIALX3375-1KG
SIALX3375-500G
SIALX3375-5G
Xylitol 99, Sigma-Aldrich®
Xylitol
Vzorec:
C₅H₁₂O₅ MW: 152.15 Bod varu: 495 Bod tání: 94 Hustota: 1.52 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064292 CAS číslo: 87-99-0 EINECS: 201-788-0 Merck Index: 13,10140 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...