Tisk…

Dichlormethan for gas chromatography

Dodavatel: SCHARLAB
Varování

Synonyms: Methylene chloride

CL03451000 CL03452500
SCARCL03451000EA 0 CZK
SCARCL03451000 SCARCL03452500
Dichlormethan for gas chromatography
Dichloromethane
Vzorec: CH₂Cl₂
MW: 84.93
Bod varu: 39.8
Bod tání: –95 °C
Hustota: 1.322
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS číslo: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR