Dichloroacetyl chloride ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B22296.22
B22296.36
B22296.22EA
1150
CZK
B22296.22
B22296.36
Dichloroacetyl chloride ≥97%
Dichloroacetyl chloride
Purity: 97%
CAS No.: 79-36-7
Molecular Formula: Cl2CHCOCl
Formula Weight: 147.39
Flash Point: None
Refractive Index: 1.4620
Boiling Point: 107-108°
Merck Reference: 14,3053
UN No.: 1765
MDL No.: MFCD00000840
Vzorec:
C₂HCl₃O Bod varu: 107…108 °C Hustota: 1,532 Bod vzplanutí: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000840 CAS číslo: 79-36-7 EINECS: 201-199-9 UN: 1765 ADR: 8,II Merck Index: 14,3053 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...