Tisk…

(±)-Isonipecotic acid 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Piperidine-4-carboxylic acid, (±)-4-Piperidinecarboxylic acid, 4-Piperidinecarboxylic acid, Isonipecotic acid

122620250 122621000 122625000
ACRO122620250EA 1310 CZK
ACRO122620250 ACRO122621000 ACRO122625000
(±)-Isonipecotic acid 99%
(±)-Isonipecotic acid
Vzorec: C₆H₁₁NO₂
Bod varu: 264 °C (1013 hPa)
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006004
CAS číslo: 498-94-2
EINECS: 207-872-3
Merck Index: 13,05208

Objednat

Výsledky specifikačního testu

Appearance White to pink-beige Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (120°C, 4 hrs)
Solubility (10 % in water) Clear to slightly hazy
Titration with HClO4 ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR