Tisk…

(±)-Nipecotic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

M-Clarity™ quality level = MQ100

211672-1G 211672-25G 211672-5G
SIAL211672-1GEA 0 CZK
SIAL211672-1G SIAL211672-25G SIAL211672-5G
(±)-Nipecotic acid, Sigma-Aldrich®
(±)-Nipecotic acid
Vzorec: C₆H₁₁NO₂
MW: 129,16 g/mol
MDL Number: MFCD00005992
CAS číslo: 498-95-3
EINECS: 207-873-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR