(±)-Nipecotic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid
B24723.03
B24723.06
B24723.14
B24723.03EA
862
CZK
B24723.03
B24723.06
B24723.14
(±)-Nipecotic acid ≥98%
(±)-Nipecotic acid
Purity: 98%
CAS No.: 498-95-3
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: ca 261° dec.
Solubility: Soluble in water
Beilstein Reference: 81096
Merck Reference: 14,6561
MDL No.: MFCD00005992
Vzorec:
C₆H₁₁NO₂ MW: 129,16 g/mol Bod tání: ∼ 261 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00005992 CAS číslo: 498-95-3 EINECS: 207-873-9 Merck Index: 14,6561 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...