(±)-Pipecolinic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(±)-2-Piperidinecarboxylic acid, DL-Pipecolinic acid, H-DL-Homopro-OH, Pipecolinic Acid, Piperidine-2-carboxylic acid, Pipecolinic acid
B23541.06
B23541.14
B23541.22
B23541.06EA
982
CZK
B23541.06
B23541.14
B23541.22
(±)-Pipecolinic acid ≥99%
(±)-Pipecolinic acid
Purity: 99%
CAS No.: 535-75-1
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: ca 280° dec.
Beilstein Reference: 81095
Merck Reference: 14,7458
MDL No.: MFCD00064347
Vzorec:
C₆H₁₁NO₂ MW: 129.16 Bod tání: ∼ 280 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00064347 CAS číslo: 535-75-1 EINECS: 208-616-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...