Ethyl-6-bromohexanoate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
6-Bromocaproic acid ethyl ester, 6-Bromohexanoic acid ethyl ester, Ethyl 6-bromohexanoate
A18398.18
A18398.09
A18398.30
A18398.18EA
3990
CZK
A18398.18
A18398.09
A18398.30
Ethyl-6-bromohexanoate ≥97%
Ethyl-6-bromohexanoate
Purity: 97+%
CAS No.: 25542-62-5
Molecular Formula: Br(CH2)5CO2CH2CH3
Formula Weight: 223.12
Flash Point:
Vzorec:
BrH₂C(CH₂)₄COOCH₂CH₃ Bod varu: 128…130 °C (16 mmHg) Bod tání: 33 °C Hustota: 1,254 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000270 CAS číslo: 25542-62-5 EINECS: 247-085-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...