Tisk…

6-Aminonicotinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid

M-Clarity™ quality level = MQ100

216879-1G 216879-5G
SIAL216879-1GEA 1530 CZK
SIAL216879-1G SIAL216879-5G
6-Aminonicotinic acid, Sigma-Aldrich®
6-Aminonicotinic acid
Vzorec: C₆H₆N₂O₂
MW: 138,13 g/mol
Bod tání: > 300 °C
MDL Number: MFCD00006326
CAS číslo: 3167-49-5
EINECS: 221-630-4
Merck Index: 13,00455

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR