Tisk…

δ-Valerolactone 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

162710500 162712500 162710010
ACRO162710500EA 4160 CZK
ACRO162710500 ACRO162712500 ACRO162710010
δ-Valerolactone 99%
δ-Valerolactone
Vzorec: C₅H₈O₂
Bod varu: 230 °C (760 mmHg)
Bod tání: –13 °C
Hustota: 1,1 g/cm³
Bod vzplanutí: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006645
CAS číslo: 542-28-9
EINECS: 208-807-1

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4590 (20°C, 589 nm)
Additional info remark: may contain variable amounts of polymerized product
GC ≥98.5 %
pH 4.5 to 6.5 (4 % in water)
Total volatile ≤5 % (including water)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR