(±)-δ-Octalactone 97
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(±)-Octan-5-olide, (±)-Tetrahydro-6-propyl-2H-pyran-2-one, δ-Octanolactone
L14303.14
L14303.22
L14303.14EA
2050
CZK
L14303.14
L14303.22
(±)-δ-Octalactone 97
(±)-δ-Octalactone
Purity: 97%
CAS No.: 698-76-0
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 125°(257°F)
Refractive Index: 1.4550
Melting Point: -14°
Boiling Point: 238°
Beilstein Reference: 111515
MDL No.: MFCD00144051
Vzorec:
C₈H₁₄O₂ MW: 142.2 Bod varu: 238 °C Bod tání: –14 °C Hustota: 1.002 Bod vzplanutí: 125 °C (257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00144051 CAS číslo: 698-76-0 EINECS: 211-820-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...