(±)-δ-Nonalactone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
W335606-1KG-K
W335606-4KG-K
SIALW335606-1KG-KEA
11150
CZK
SIALW335606-1KG-K
SIALW335606-4KG-K
(±)-δ-Nonalactone, Sigma-Aldrich®
(±)-δ-Nonalactone
Vzorec:
C₉H₁₆O₂ |
MDL Number:
MFCD00036500 CAS číslo: 3301-94-8 EINECS: 221-974-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...