Tisk…

(±)-δ-Hexalactone ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: (±)-Hexan-5-olide, (±)-Tetrahydro-6-methyl-2H-pyran-2-one, 6-Methyltetrahydro-2H-pyran-2-one, (±)-δ-Caprolactone

A19274.14 A19274.22
A19274.14EA 1840 CZK
A19274.14 A19274.22
(±)-δ-Hexalactone ≥98%
(±)-δ-Hexalactone

Purity: 98% CAS No.: 823-22-3 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 105°(221°F) Refractive Index: 1.4510 Melting Point: 18° Boiling Point: 231° Beilstein Reference: 80501 MDL No.: MFCD00083574

Vzorec: C₆H₁₀O₂
Bod varu: 231 °C
Bod tání: 18 °C
Hustota: 1,040
Bod vzplanutí: 105 °C (221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00083574
CAS číslo: 823-22-3
EINECS: 212-511-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR