(±)-δ-Dodecalactone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
6-Heptyltetrahydro-2H-pyran-2-one, δ-Dodecalactone, 5-Dodecanolide, δ-Dodecanolactone, (±)-Tetrahydro-6-heptyl-2H-pyran-2-one, (±)-Dodecan-5-olide
L14406.14
L14406.22
L14406.14EA
1240
CZK
L14406.14
L14406.22
(±)-δ-Dodecalactone ≥98%
(±)-δ-Dodecalactone
Purity: 98%
CAS No.: 713-95-1
Molecular Formula: C12H22O2
Formula Weight: 198.30
Flash Point: 165°(329°F)
Refractive Index: 1.4600
Melting Point: -7°
Boiling Point: 288°
Beilstein Reference: 1282749
MDL No.: MFCD00006651
Vzorec:
C₁₂H₂₂O₂ MW: 198.31 Bod varu: 288 °C Bod tání: –7 °C Hustota: 0.950 Bod vzplanutí: 165 °C (329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006651 CAS číslo: 713-95-1 EINECS: 211-932-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...