Tisk…

(±)-δ-Dodecalactone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Heptyltetrahydro-2H-pyran-2-one, δ-Dodecalactone, 5-Dodecanolide, δ-Dodecanolactone, (±)-Tetrahydro-6-heptyl-2H-pyran-2-one, (±)-Dodecan-5-olide

M-Clarity™ quality level = MQ100

W240109-10KG-K W240109-1KG-K W240109-5KG-K
SIALW240109-10KG-KEA 28620 CZK
SIALW240109-10KG-K SIALW240109-1KG-K SIALW240109-5KG-K
(±)-δ-Dodecalactone, Sigma-Aldrich®
(±)-δ-Dodecalactone
Vzorec: C₁₂H₂₂O₂
MW: 198,31 g/mol
Bod varu: 126…128 °C (0,5 mmHg)
Bod tání: –7 °C
Hustota: 0,9585 g/cm³ (20 °C)
MDL Number: MFCD00006651
CAS číslo: 713-95-1
EINECS: 211-932-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR