Tisk…

(±)-γ-Undecalactone ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (±)-Undecan-4-olide, Gamma-undecalactone, Undecanoic gamma-Lactone

L16028.AE L16028.AP
L16028.AEEA 1630 CZK
L16028.AE L16028.AP
(±)-γ-Undecalactone ≥99%
(±)-γ-Undecalactone

Purity: 99% CAS No.: 104-67-6 Molecular Formula: C11H20O2 Formula Weight: 184.28 Flash Point: 156°(312°F) Refractive Index: 1.4500 Boiling Point: 164-166°/13mm Beilstein Reference: 81943 MDL No.: MFCD00005405

Vzorec: C₁₁H₂₀O₂
MW: 184.28
Bod varu: 164…166 °C (13 mmHg)
Hustota: 0.9414
Bod vzplanutí: 156 °C (312 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005405
CAS číslo: 104-67-6
EINECS: 203-225-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR