(±)-γ-Hexalactone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(±)-γ-Caprolactone, γ-Caprolactone, 5-Ethyldihydrofuran-2(3H)-one, (±)-Hexan-4-olide
Purity: 98%
CAS No.: 695-06-7
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 98°(208°F)
Refractive Index: 1.4390
Melting Point: -18°
Boiling Point: 218-219°
Beilstein Reference: 107260
MDL No.: MFCD00005401
Vzorec:
C₆H₁₀O₂ MW: 114,14 g/mol Bod varu: 218…219 °C Bod tání: –18 °C Hustota: 1,025 Bod vzplanutí: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005401 CAS číslo: 695-06-7 EINECS: 211-778-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...